Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, Naphthalene 5,000 µg/ml in methanol, Supelco®
Organic reference standard, Naphthalene 5,000 µg/ml in methanol, Supelco®
Catalog # SUPL40053
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Naphthalene 5,000 µg/ml in methanol, Supelco®
Catalog # SUPL40053
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    91-20-3
  • Description:
    Naphthalene 5,000 µg/ml in methanol
  • Formula:
    C₁₀H₈
  • Description:
    Naphthalene

Specifications

About this item