Sulphuric acid fuming (20% free SO₃) (active SO3), Puriss. p.a. analytical reagent
30736-1L
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
- Residue on ignition:Max. 0.01 %
- Ammonium (NH₄):Max. 0.0002 %
- Arsenic (As):Max. 0.000003 %
- Iron (Fe):Max. 0.005 %
- Heavy metals (as Pb):Max. 0.0005 %
- Chloride (Cl):Max. 0.0005 %
- Total SO₃:Max. 87 %
- Free SO₃:20 - 30 %
- Vapor Density:3 (vs air)
: Oleum
:
:80.06
:1831
:GHS07
:GHS05
:GHS08
:Ambient
:8014-95-7
:H₂O₇S₂ (H₂SO₄ + SO₃)
:MFCD00137792
:–11 °C
:1.90 g/cm³ (20 °C)
:8 I