Order Entry
Netherlands
ContactUsLinkComponent
Sodium hypochlorite (5 - 6% Cl₂) in aqueous solution
 
 
Sodium hypochlorite (5 - 6% Cl₂) in aqueous solution
  VWRS9851.5000
 :  
 
 
Sodium hypochlorite (5 - 6% Cl₂) in aqueous solution
  VWRS9851.5000
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

 

 

 :
 :74.44 g/mol
 :111 °C (1013 hPa)
 :–30…–20 °C
 :1.22…1.25 g/cm³ (20 °C)
 :Refrigerator
 :231-668-3
 :1791
 :8,II
 :GHS05
 :GHS09
 :7681-52-9
 :ClNaO
 :MFCD00011120