Order Entry
Netherlands
ContactUsLinkComponent
Orthoboric acid 40 g/l in aqueous solution for Kjeldahl determinations, with indicator
Orthoboric acid 40 g/l in aqueous solution for Kjeldahl determinations, with indicator
Catalog # 192316H
Supplier:  VWR Chemicals
CAS Number:  
undefined
Orthoboric acid 40 g/l in aqueous solution for Kjeldahl determinations, with indicator
Catalog # 192316H
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    39.0 - 41.0 g/l
  • pH (20°C):
    3.80 - 4.0

Specification Test Results

About this item

 :
 :61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337