Order Entry
Netherlands
ContactUsLinkComponent
Human Recombinant SCF (Animal Free) (from E. coli)
Human Recombinant SCF (Animal Free) (from E. coli)
Certificates
undefined
Human Recombinant SCF (Animal Free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6,7×10⁴ units/mg
  • Gene ID:
    P21583
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    c-Kit Ligand|KL|MGF|Steel Factor
  • Protein/peptide name:
    SCF
  • Purity:
    ≥95%
  • Molecular weight:
    18,6 kDa
  • Sequence:
    MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together