Order Entry
Netherlands
ContactUsLinkComponent
 
Octadecane, standard material for GC
Catalog # TCIAS0290-5ML
Supplier:  TCI
CAS Number:  
undefined
Octadecane, standard material for GC
Catalog # TCIAS0290-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • CAS No.:
    593-45-3
  • Description:
    Octadecane, Purity (GC): min. 99.5 %
  • Environmentally Preferable:
  • Formula:
    H₃C(CH₂)₁₆CH₃
  • Description:
    Octadecane
  • Appearance:
    White to Almost white powder to lump

Specifications

About this item