Lithium pyruvate monohydrate, guaranteed for standard to GOT, GPT
TCIAP0659-25G
: TCI
:
- Pk:25 g
- CAS No.:2922-61-4
- Description:Lithium Pyruvate Monohydrate, Purity (Iodometric Titration): min. 95.0 %(calcd.on anh.substance)
- Formula:C₃H₃LiO₃.H₂O
- Storage temperature:Refrigerator
- Description:Lithium Pyruvate Monohydrate
- Appearance:White powder to crystal