1-Heptene, Standard Material for GC
Catalog # TCIAS0337-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- CAS No.:592-76-7
- Description:1-Heptene, Purity (GC): min. 99.5 %
- Formula:C₇H₁₄
- Description:1-Heptene
- Flash point:–4 °C (lit.)
- Boiling point:94 °C (lit.)
- Appearance:Colorless clear liquid