Order Entry
Netherlands
ContactUsLinkComponent
2,6-Dinitro-p-cresol ≥98.0% (by titrimetric analysis) wetted with ca. 20% water (unit weight on dry weight basis)
Toxic
2,6-Dinitro-p-cresol ≥98.0% (by titrimetric analysis) wetted with ca. 20% water (unit weight on dry weight basis)
Catalog # TCIAD2166-25G
Supplier:  TCI
CAS Number:  
Toxic
2,6-Dinitro-p-cresol ≥98.0% (by titrimetric analysis) wetted with ca. 20% water (unit weight on dry weight basis)
Catalog # TCIAD2166-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    78.0 to 82.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :198.14 g/mol
Melting Pt:78…82 °C
EINECS:210-203-8
UN:2811
ADR:6.1,II
 :GHS06
CAS Number:609-93-8
Formula:C₇H₆N₂O₅
MDL Number:MFCD00007121