Dodecyl 2-nitrophenyl ether ≥99.0%, Selectophore™ ionophore, Supelco®
Catalog # 44224-5ML
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:307.43 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:83027-71-8
Formula:C₁₈H₂₉NO₃
MDL Number:MFCD00024221