Chromoionophore III, Selectophore™ ionophore, Supelco®
Catalog # 27088-10MG-F
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:569.86 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:149683-18-1
Formula:C₃₈H₅₅N₃O
MDL Number:MFCD00213937