Order Entry
Netherlands
ContactUsLinkComponent
n-Decane ≥95%, GPR RECTAPUR®
n-Decane ≥95%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
n-Decane ≥95%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 95 %
  • Boiling point:
    Min. 170 °C
  • Density (20/4):
    0,729 --> 0,731
  • Aromatics:
    Max. 100 ppm

Specification Test Results

Frequently Bought Together