Order Entry
Netherlands
ContactUsLinkComponent
 
Cefixime trihydrate ≥98.0% (by HPLC) antibiotic, for microbiology
  18588-100MG-F
 :  Merck
 :  
Cefixime trihydrate ≥98.0% (by HPLC) antibiotic, for microbiology
  18588-100MG-F
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

 :
 :507.489 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS08
 :125110-14-7
 :C₁₆H₂₁N₅O₁₀S₂
 :MFCD03788802