Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
TPPS hydrate (=tetraphenylporphyrin tetrasulphonic acid hydrate) ≥95.0% for spectrophotometric determination of transition metals, Supelco®
Catalog # 89456-100MG-F
Supplier:  Merck
CAS Number:  
undefined
Irritant
TPPS hydrate (=tetraphenylporphyrin tetrasulphonic acid hydrate) ≥95.0% for spectrophotometric determination of transition metals, Supelco®
Catalog # 89456-100MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :934.98 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
CAS Number:35218-75-8
Formula:C₄₄H₃₀N₄O₁₂S₄
MDL Number:MFCD00070632