Order Entry
Netherlands
ContactUsLinkComponent
Dibutylammonium acetate 0.5 M in water, LiChropur™ for ion pair chromatography, Supelco®
Dibutylammonium acetate 0.5 M in water, LiChropur™ for ion pair chromatography, Supelco®
Catalog # 73345-6EA
Supplier:  Merck
CAS Number:  
undefined
Dibutylammonium acetate 0.5 M in water, LiChropur™ for ion pair chromatography, Supelco®
Catalog # 73345-6EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :189.30 g/mol
CAS Number:19070-91-8
Formula:C₈H₁₉N
MDL Number:MFCD00191381