Sodium carbonate : Sodium hydroxide concentrate (150 mM : 15 mM) in water IC eluent concentrate (20x) for Metrosep A Supp 16, Supelco®
: Merck
Sodium carbonate : Sodium hydroxide concentrate (150 mM : 15 mM) in water IC eluent concentrate (20x) for Metrosep A Supp 16, Supelco®
: Merck
:
Frequently Bought Together
Sodium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Bottles, narrow or wide neck, soda-lime glass, clear or amber, with standard ground joint
: GLASWARENFABRIK KARL HECHT
Ammonium hydrogen difluoride ≥98%, GPR RECTAPUR®
: VWR Chemicals