Bestelling invoeren
Netherlands
ContactUsLinkComponent
Pharmaceutical secondary standard, Fluoxetine Hydrochloride, certified reference material, Supelco®
Pharmaceutical secondary standard, Fluoxetine Hydrochloride, certified reference material, Supelco®
Catalog # PHR1394-1G
Supplier:  Merck
CAS Number:  
undefined
Pharmaceutical secondary standard, Fluoxetine Hydrochloride, certified reference material, Supelco®
Catalog # PHR1394-1G
Supplier:  Merck
CAS Number:  
Specifications
  • Concentratie:
    Neat
  • VPE:
    1 g
  • Standaard:
    Pharmaceutical secondary standard (CRM)
  • CAS No.:
    56296-78-7
  • Omschrijving:
    Fluoxetine Hydrochloride
  • Omschrijving:
    Fluoxetine Hydrochloride
Specifications
About this item
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.