Supelco® Organic Reference Standard, ᴅ-Pantethine
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:16816-67-4
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD00151211
- M-Clarity quality level:MQ100
- Synonym(s):Bis(N-pantothenylamidoethyl) disulfide; LBF factor; Pantethein oxidized form
- Molecular weight:554,72
Specifications
Frequently Bought Together