Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, Dopamine-1,1,2,2-d₄ hydrochloride
Supelco® Organic Reference Standard, Dopamine-1,1,2,2-d₄ hydrochloride
Catalogus # 73483-5MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Dopamine-1,1,2,2-d₄ hydrochloride
Catalogus # 73483-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    203633-19-6
  • Description:
    Dopamine-1,1,2,2-d₄ hydrochloride
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01317352
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-(3,4-Dihydroxyphenyl)ethyl-1,1,2,2-d₄-amine hydrochloride
  • Melting point:
    248 - 250 °C (lit.)
  • Formula:
    C₈D₄H₇O₂N.HCl
  • Molecular weight:
    193,66

Specifications

About this item

  • GHS signal: GHS05, GHS07