Supelco® Organic Reference Standard, Theaflavin monogallate
Catalog # 53963-1MG
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mg
- Standards:Analytical standard
- CAS No.:31629-79-5
- Description:Theaflavin monogallate
- Storage conditions:–20 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Molecular weight:716,60