Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
2-NP-AOZ-d4, analytical standard, VETRANAL™, Supelco®
2-NP-AOZ-d4, analytical standard, VETRANAL™, Supelco®
34008-10MG-R
:
Merck
:
2-NP-AOZ-d4, analytical standard, VETRANAL™, Supelco®
34008-10MG-R
:
Merck
:
Pk:
10 mg
Standards:
analytical standard
CAS No.:
1007478-57-0
Description:
2-NP-AOZ-d4
M-Clarity quality level:
MQ100
Description:
2-NP-AOZ-d4