Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Cymiazole
Catalog # 32407-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Cymiazole
Catalog # 32407-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    61676-87-7
  • Description:
    Cymiazole
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01311800
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2,4-Dimethyl-N-(3-methyl-2(3H)-thiazolylidene)benzenamine; 2-(2,4-Dimethylphenylimino)-3-methylthiazoline; 2-(2,4-Xylylimino)-3-methylthiazoline; N-[(2EZ)-3-Methyl-1,3-thiazol-2-(3H)ylidene]-2,4-xylidine
  • Molecular weight:
    218,32
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501