


Chloramine-T (sodium salt) trihydrate 98.0-103%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS
Catalog # 22686.230
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Assay:98.0 - 103.0 %
- Identification A:Passes test Reag.Ph.Eur.
- Identification B:Passes test Reag.Ph.Eur.
- Identification C:Passes test Reag.Ph.Eur.
- Identification D:Passes test Reag.Ph.Eur.
- Identification E:Passes test Reag.Ph.Eur.
- Appearance of solution:Passes test Reag.Ph.Eur.
- Clarity of aqueous solution:Passes test ACS
- Ortho compounds:Passes test
- Suitability for determination of bromide:Passes test ACS
- pH (20°C; 5 %):8.0 - 10.0
- pH (25°C; 5 %):8.0 - 10.0
- Residue insoluble in anhydrous ethanol:Max. 1.5 %
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
About this item
:
ADR:8,III
:281.69 g/mol
UN:3263
:GHS07
:GHS05
:GHS08
CAS Number:7080-50-4
Formula:CH₃C₆H₄SO₂NClNa·3H₂O
MDL Number:MFCD00149066