Supelco® Organic Reference Standard, trans-Cinnamic acid
Catalog # 97013-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:140-10-3
- Description:trans-Cinnamic acid
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD00004369
- M-Clarity quality level:MQ100
- Synonym(s):(2E)-3-Phenyl-2-propenoic acid; trans-3-Phenylacrylic acid
- Melting point:133 °C (lit.); 133 - 137 °C
- Molecular weight:148,16
- Flash point:160 °C