Organic reference standard, Sodium nitrate, analytical standard, Supelco®
Catalog # 15736-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Standards:Analytical standard
- CAS No.:7631-99-4
- Description:Sodium nitrate, ≥99,995% trace metals basis
- Melting point:306 °C
- Formula:NaNO₃
- Description:Sodium nitrate
- Form:Neat