Supelco® Organic Reference Standard, Propionyl-ʟ-carnitine-(N-methyl-d₃)
52941-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:203806-01-3
- Description:Propionyl-ʟ-carnitine-(N-methyl-d₃)
- Seq:03
- Storage conditions:–20 °C
- Shipping conditions:WICE
- MDL No.:MFCD01073492
- M-Clarity quality level:MQ100
- Synonym(s):(2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d₃)-2-(1-oxopropoxy)-1-propanaminium inner salt; ʟ-Carnitine-(methyl-d₃) propionyl ester; Propanoyl-ʟ-carnitine-(N-methyl-d₃)
- Molecular weight:220,28
- GHS signal: GHS07