Supelco® Organic Reference Standard, Neohesperidin dihydrochalcone
Catalog # 75041-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:20702-77-6
- Description:Neohesperidin dihydrochalcone
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00017711
- M-Clarity quality level:MQ100
- Synonym(s):1-(4-((2-O-[6-Deoxy-α-ʟ-mannopyranosyl]-β-ᴅ-glucopyranosyl)oxy)-2,6-dihydroxyphenyl)-3-[3-hydroxy-4-methoxyphenyl]-1-propanone; NHDC
- Melting point:156 - 158 °C (lit.)
- Molecular weight:612,58
Specifications
About this item
Caution: P264 - P270 - P301 + P312 - P501