Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, N-(2-Methylbutyryl)Glycine
Supelco® Organic Reference Standard, N-(2-Methylbutyryl)Glycine
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, N-(2-Methylbutyryl)Glycine
Supplier:  Merck

Specifications

  • CAS No.:
    52320-67-9
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H315 - H319
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD09736911
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-Methylbutyryl glycine; N-(2-Methyl-1-oxobutyl)glycine
  • Formula:
    C₇H₁₃NO₃
  • Molecular weight:
    159,18
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

Specifications

Frequently Bought Together