Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, ʟ-Methionine hydrochloride solution
Supelco® Organic Reference Standard, ʟ-Methionine hydrochloride solution
  50272-5ML-F
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, ʟ-Methionine hydrochloride solution
  50272-5ML-F
 :  Merck
 :  

 

  • Concentration:
    100 mM amino acid in 0,1 M HCl
  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    6810-12-4
  • Description:
    ʟ-Methionine hydrochloride solution
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD03791082
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    185,67