Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, ʟ-Cystine hydrochloride solution
Supelco® Organic Reference Standard, ʟ-Cystine hydrochloride solution
Catalog # 57579-5ML-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, ʟ-Cystine hydrochloride solution
Catalog # 57579-5ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 mM amino acid in 0,1 M HCl
  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    34760-60-6
  • Description:
    ʟ-Cystine hydrochloride solution
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H290
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00068399
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    276,76

Specifications

About this item

Caution: P234 - P390