Supelco® Organic Reference Standard, ʟ-Arginine Hydrochloride Solution
08163-5ML-F
: Merck
:
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:1119-34-2
- Description:ʟ-Arginine hydrochloride solution
- Storage conditions:2...8 °C
- Hazard class description:H290
- Shipping conditions:STD
- MDL No.:MFCD00064550
- Formula:C₆H₁₄N₄O₂ ·HCl
- Molecular weight:210,66
- Storage temperature:2...8 °C
- Flash point:Not applicable
: P234 - P390