Organic reference standard, Kojic acid, analytical standard, Supelco®
95197-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:501-30-4
- Description:Kojic acid, ≥99,0% (HPLC)
- Melting point:152...155 °C
- Formula:C₆H₆O₄
- Description:Kojic acid
- Form:Neat
Tyrosinase inhibitor.