Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Itaconic Acid
  93598-100MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Itaconic Acid
  93598-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    97-65-4
  • Description:
    Itaconic acid
  • Storage conditions:
    RT
  • Hazard class description:
    H318
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00004260
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Methylenesuccinic acid
  • Melting point:
    165...168 °C (lit.)
  • Formula:
    HO₂CCH₂C(=CH₂)CO₂H
  • Molecular weight:
    130,10
  • Density:
    1,573 g/ml at 25 °C (lit.)
  • Form:
    Neat

 

 

  • GHS signal: GHS07
 : P280 - P305 + P351 + P338 + P310