Supelco® Organic Reference Standard, Ginkgolic acid C13:0
Catalogus # 49962-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:20261-38-5
- Description:Ginkgolic acid C13:0
- Storage conditions:2…8 °C
- Hazard class description:H317
- Shipping conditions:STD
- MDL No.:MFCD01661602
- M-Clarity quality level:MQ100
- Synonym(s):2-Hydroxy-6-tridecylbenzoic acid; 6-Tridecylsalicylic acid; Ginkgoneolic acid
- Molecular weight:320,47
Specifications
About this item
- GHS signal: GHS07
Caution: P280 - P302 + P352