Organic reference standard, gamma-Hexalactone, analytical standard, Supelco®
Catalog # 68554-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:695-06-7
- Description:gamma-Hexalactone, ≥99,0% (GC)
- Formula:C₆H₁₀O₂
- Description:gamma-Hexalactone
- Boiling point:219 °C (lit.)