Supelco® Organic Reference Standard, ᴅ-(+)-Raffinose pentahydrate
95068-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:17629-30-0
- Description:ᴅ-(+)-Raffinose pentahydrate
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00071590
- M-Clarity quality level:MQ100
- Synonym(s):O-α-ᴅ-Galactopyranosyl-(1→6)-α-ᴅ-glucopyranosyl β-ᴅ-fructofuranoside; Melitose; Melitriose
- Molecular weight:594,51