Organic reference standard, cis-1,3-Pentadiene, analytical standard, Supelco®
Catalog # 09207-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:1574-41-0
- Description:cis-1,3-Pentadiene, ≥97,0% (GC)
- Melting point:−141 °C (lit.)
- Formula:C₅H₈
- Storage temperature:2...8 °C
- Description:cis-1,3-Pentadiene
- Boiling point:44 °C(lit.)