Organic reference standard, beta-Zearalenol solution, analytical standard, Supelco®
Catalog # 35409-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:10 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:71030-11-0
- Description:beta-Zearalenol solution
- Formula:C₁₈H₂₄O₅
- Storage temperature:−20 °C
- Description:beta-Zearalenol solution