Supelco® Organic Reference Standard, 2-Isopropyl-D₇-Thioxanthen-9-One
Catalog # 33652-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:1173019-24-3
- Description:2-Isopropyl-d₇-thioxanthen-9-one
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD09842833
- M-Clarity quality level:MQ100
- Synonym(s):ITX-d₇
- Formula:C₁₆D₇H₇OS
- Molecular weight:261,39
- Flash point:Not applicable
- Form:Neat