Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, (E)-3-Methylglutaconic Acid
Supelco® Organic Reference Standard, (E)-3-Methylglutaconic Acid
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, (E)-3-Methylglutaconic Acid
Supplier:  Merck

Specifications

  • CAS No.:
    372-42-9
  • Melting point:
    137...143 °C
  • Formula:
    C₆H₈O₄
  • Storage temperature:
    2…8 °C
  • Description:
    (E)-3-Methylglutaconic acid
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H315 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01556044
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (2E)-3-Methyl-2-pentenedioic acid
  • Molecular weight:
    144,13
  • Flash point:
    Not applicable

Specifications

Frequently Bought Together