Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, (+)-trans-Limonene 1,2-epoxide
Supelco® Organic Reference Standard, (+)-trans-Limonene 1,2-epoxide
Catalog # 62136-1ML-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, (+)-trans-Limonene 1,2-epoxide
Catalog # 62136-1ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    6909-30-4
  • Description:
    (+)-trans-Limonene 1,2-epoxide
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD16652609
  • Synonym(s):
    (+)-trans-p-Mentha-1,8-diene 1,2-epoxide; (1S,2R,4R)-4-Isopropenyl-1-methyl-1-cyclohexene 1,2-epoxide; (1S,4R)-1,2-Epoxy-p-menth-8-ene
  • Molecular weight:
    152,23
  • Density:
    0,930 g/ml at 20 °C (lit.)
  • Flash point:
    65 °C - Closed cup

Specifications

About this item