Order Entry
Netherlands
ContactUsLinkComponent
Pesticide standard, Spirodiclofen, reference material, Honeywell Fluka™
Pesticide standard, Spirodiclofen, reference material, Honeywell Fluka™
Catalog # HONCR1642-100MG
CAS Number:  
Pesticide standard, Spirodiclofen, reference material, Honeywell Fluka™
Catalog # HONCR1642-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    148477-71-8
  • Description:
    Spirodiclofen
  • Formula:
    C₂₁H₂₄Cl₂O₄
  • Storage temperature:
    Ambient
  • Description:
    Spirodiclofen
  • Form:
    Powder

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.