Order Entry
Netherlands
ContactUsLinkComponent
Sealing film, self-adhesive for PCR, real time PCR
Catalog # 391-0705
Supplier:  Brand
undefined
Sealing film, self-adhesive for PCR, real time PCR
Catalog # 391-0705
Supplier:  Brand
Specifications
  • Packed:
    50/pack (each 8 strips)
  • Description:
    Sealing film, self-adhesive, for PCR, Real-time PCR
  • Material:
    Polyester
  • Pk:
    400
Specifications
About this item
Polyester flims suitable for PCR and Real-Time PCR measurements. Temperature range: –40 °C to +120 °C.