Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Valifenalate
Catalog # 37078-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Valifenalate
Catalog # 37078-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    PESTANAL®, Analytical standard, mixture of isomers
  • CAS No.:
    283159-90-0
  • Description:
    Valifenalate
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD22373630
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N-[(1-Methylethoxy)carbonyl]-ʟ-valyl-3-(4-chlorophenyl)-β-alanine methyl ester; Methyl N-(isopropoxycarbonyl)-ʟ-valyl-(3RS)-3-(4-chlorophenyl)-β-alaninate
  • Formula:
    C₁₉H₂₇ClN₂O₅
  • Molecular weight:
    398,88
  • Flash point:
    Not applicable

Specifications

About this item