Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Tri-o-tolyl phosphate
Catalog # 51885-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Tri-o-tolyl phosphate
Catalog # 51885-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    78-30-8
  • Description:
    Tri-o-tolyl phosphate
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 - H370 - H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00041906
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    NSC 438; Phosphoric acid tris(2-methylphenyl) ester; TOTP; Tri-o-cresyl phosphate
  • Molecular weight:
    368,36
  • Flash point:
    110 °C

Specifications

About this item

  • GHS signal: GHS08, GHS07, GHS09
Caution: P260 - P264 - P270 - P273 - P301 + P312 - P308 + P311