Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, Tricosafluorododecanoic acid
Supelco® Organic Reference Standard, Tricosafluorododecanoic acid
Catalog # 92291-50MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Tricosafluorododecanoic acid
Catalog # 92291-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    307-55-1
  • Description:
    Tricosafluorododecanoic acid
  • Storage conditions:
    RT
  • Hazard class description:
    H302 + H332 - H351 - H360D - H362 - H372
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00198081
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    PFAS; Perfluorododecanoic acid; Perfluorolauric acid
  • Melting point:
    105 - 108 °C (lit.)
  • Formula:
    CF₃(CF₂)₁₀CO₂H
  • Molecular weight:
    614,10
  • Boiling point:
    245 °C/740 mmHg (lit.)

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P202 - P260 - P263 - P301 + P312 - P304 + P340 + P312 - P308 + P313