Supelco® PESTANAL® Organic Reference Standard, Triclopyr 2-Butoxyethylester
Catalog # 36538-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:64700-56-7
- Description:Triclopyr 2-butoxyethylester
- Storage conditions:2...30 °C
- Hazard class description:H302 - H315 - H400
- Shipping conditions:STD
- MDL No.:MFCD00145446
- M-Clarity quality level:MQ100
- Formula:C₁₃H₁₆Cl₃NO₄
- Molecular weight:356,63
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P264 - P273 - P280 - P301 + P312 - P302 + P352 - P332 + P313