Organic reference standard, trans-Nonachlor, analytical standard, Supelco®
Catalog # 442811.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Analytical standard
- CAS No.:39765-80-5
- Description:trans-Nonachlor
- Storage temperature:RT
- Description:trans-Nonachlor
- Pk Info:25 mg/ampule