Supelco® Organic Reference Standard, trans-9,12-Octadecadienoic acid methyl ester
Catalog # 46951-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:10 mg/ml in heptane
- Pk:1
- Standards:Certified reference material
- CAS No.:2566-97-4
- Description:trans-9,12-Octadecadienoic acid methyl ester
- Environmentally Preferable:
- Storage conditions:–10...–25 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:–4 °C
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331