Order Entry
Netherlands
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Thidiazuron
Supelco® PESTANAL® Organic Reference Standard, Thidiazuron
  45686-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Thidiazuron
  45686-250MG
 :  Merck
 :  

 

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    51707-55-2
  • Description:
    Thidiazuron
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H315 - H319 - H335 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078723
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₉H₈N₄OS
  • Molecular weight:
    220,25
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07, GHS09
 : P261 - P264 - P271 - P273 - P302 + P352 - P305 + P351 + P338